missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adapalene 98.0+%, TCI America™

$205.40 - $718.90

Chemical Identifiers

CAS 106685-40-9
Molecular Formula C28H28O3
Molecular Weight (g/mol) 412.529
MDL Number MFCD03106112
InChI Key LZCDAPDGXCYOEH-UHFFFAOYSA-N
Synonym adapalene, differin, adapalenum, adapaleno, adapaleno inn-spanish, adaferin, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adapalenum inn-latin, differine, 6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid
PubChem CID 60164
ChEBI CHEBI:31174
IUPAC Name 6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
SMILES COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A2549200MG
View Documents
TCI America
A2549200MG
200 mg
Each for $205.40
 
A25491G
View Documents
TCI America
A25491G
1 g
Each for $718.90
 
Description

Description

Specifications

Chemical Identifiers

106685-40-9
412.529
LZCDAPDGXCYOEH-UHFFFAOYSA-N
60164
6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
C28H28O3
MFCD03106112
adapalene, differin, adapalenum, adapaleno, adapaleno inn-spanish, adaferin, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adapalenum inn-latin, differine, 6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid
CHEBI:31174
COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5

Specifications

106685-40-9
White-Yellow
MFCD03106112
adapalene, differin, adapalenum, adapaleno, adapaleno inn-spanish, adaferin, 6-3-adamantan-1-yl-4-methoxyphenyl-2-naphthoic acid, adapalenum inn-latin, differine, 6-3-1-adamantyl-4-methoxyphenyl-2-naphthoic acid
COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)C5
412.529
CHEBI:31174
≥98.0% (HPLC,T)
Adapalene
324°C
C28H28O3
200 mg
LZCDAPDGXCYOEH-UHFFFAOYSA-N
6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
60164
412.53
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : QJ1987000

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'