Eppendorf™ Adapter for S-4x400 Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
$534.51 - $593.25
Specifications
For Use With (Equipment) S-4x400 Bucket
No. per Pack 2/Pk.
View More Specs
Products 11
Catalog Number Mfr. No. Capacity Price Quantity  
Catalog Number Mfr. No. Capacity Price Quantity  
13864443
View Documents Promotion Details
Eppendorf™
5910714009
1 x 175 to 225 mL Conical Tubes
Each for $593.25
13864437
View Documents Promotion Details
Eppendorf™
5910705000
1 x 175 to 250 mL Flat/Conical Bottle
Each for $589.59
05414347
View Documents Promotion Details
Eppendorf™
5910706006
1 x 400 mL Bottle
Pack of 2 Each for $534.51
13864442
View Documents Promotion Details
Eppendorf™
5910712006
50 mL Self-standing Tubes, 50 mL Centriprep, 30 mL Oak Ridge
Each for $593.25
13864433
View Documents Promotion Details
Eppendorf™
5910701004
4 x 50 mL Conical Tubes
Each for $589.59
13864434
View Documents Promotion Details
Eppendorf™
5910702000
7 x 5.0 mL Microtubes, 15 mL Conical Tubes
Each for $589.59
13864439
View Documents Promotion Details
Eppendorf™
5910709005
8 x 9 mL Round Bottom Tubes
Each for $593.25
13864436
View Documents Promotion Details
Eppendorf™
5910704003
11 x 7.5 to 12 mL Round Bottom Tubes
Each for $593.25
13864435
View Documents Promotion Details
Eppendorf™
5910703007
15 x 4 to 8 mL Round Bottom Tubes
Each for $589.59
13864441
View Documents Promotion Details
Eppendorf™
5910711000
17 x 12 x 75 mm Round Bottom Tubes
Each for $593.25
13864438
View Documents Promotion Details
Eppendorf™
5910708009
26 x 1.5/2.0 mL Microtubes
Each for $593.25
Description
Description
  • Suitable for S-4x400 Rotor buckets
  • Accommodates selected array of tubes or bottles
Specifications
Specifications
S-4x400 Bucket
2/Pk.
SDS
missing translation for 'documents'
missing translation for 'documents'