missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Bottle Top Dispenser Adapter

Manufactured with PP material and can withstand fumic reagents. This Bottle Top Dispenser Adapter is durable and light weight.

$126.03 - $126.03

Specifications

Product Type Adapter
Certifications/Compliance CE Certificate, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655, Calibration lab certification by NABL as per ISO 17025, Above certificates applicable for the whole product
Color Translucent
Material Polypropylene
For Use With (Equipment) Bottletop Dispenser
View More Specs

Products 3
Catalog Number Mfr. No. Model Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Model Diameter (Metric) Price Quantity  
13681520
View Documents Promotion Details
N/A ADPT-36 36 mm
Each for $126.03
 
13681521
View Documents Promotion Details
N/A ADPT-40 40 mm
Each for $126.03
 
13681522
View Documents Promotion Details
N/A ADPT-45 45 mm
Each for $126.03
 
Description

Description

  • High quality, durable 
  • Manufactured with PP material; can withstand fumic reagents
  • Very light weight
  • Available with inlet diameters of 36, 40, and 45 mm
 
Specifications

Specifications

Adapter
Translucent
Bottletop Dispenser
CE Certificate, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655, Calibration lab certification by NABL as per ISO 17025, Above certificates applicable for the whole product
Polypropylene
Round
SDS
missing translation for 'documents'

missing translation for 'documents'