missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Adenine, 99%

Supplier:  Thermo Scientific Chemicals A1490614

 View more versions of this product

Catalog No. AAA1490614


Explore available promotions

Description

Description

Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

Solubility
Soluble in hot water, methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

Adenine
99%
MFCD00041790
UN2811
14,152
GFFGJBXGBJISGV-UHFFFAOYSA-N
7H-purin-6-amine
190
135.13
73-24-5
C5H5N5
25 g
5777
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
NC1=C2NC=NC2=NC=N1
135.13
CHEBI:16708
99%

Chemical Identifiers

73-24-5
135.13
GFFGJBXGBJISGV-UHFFFAOYSA-N
190
7H-purin-6-amine
C5H5N5
MFCD00041790
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
CHEBI:16708
NC1=C2NC=NC2=NC=N1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301

GHS P Statement
P264-P270-P301+P310-P321-P330-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : AU6125000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only