missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adenine Hemisulfate Dihydrate, ≥99%, MP Biomedicals™

A purine nucleobase with a wide range of chemical and biochemical roles in vivo and in vitro; It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP)

Supplier:  MP Biomedicals Inc 0210019501

 View more versions of this product

Catalog No. ICN10019501



Description

Description

Specifications

Specifications

Adenine Hemisulfate Dihydrate
White
C10H16N10O6S
1 g
MKPCNMXYTMQZBE-UHFFFAOYSA-N
bis(7H-purin-6-amine) sulfuric acid dihydrate
44134512
Solid
6509-19-9
≥99%
MFCD00150149
9h-purin-6-amine hemisulfate dihydrate, adenine hemisulfate dihydrate, 6-aminopurine sulfate dihydrate, bis ade sulfuric acid dihydrate, sulfuric acid bis vitamin b4 dihydrate, bis adenine sulfuric acid dihydrate, sulfuric acid-7h-purin-6-amine-water 1/2/2
O.O.OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
404.36
404.37

Chemical Identifiers

6509-19-9
404.36
MKPCNMXYTMQZBE-UHFFFAOYSA-N
44134512
O.O.OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
C10H16N10O6S
MFCD00150149
9h-purin-6-amine hemisulfate dihydrate, adenine hemisulfate dihydrate, 6-aminopurine sulfate dihydrate, bis ade sulfuric acid dihydrate, sulfuric acid bis vitamin b4 dihydrate, bis adenine sulfuric acid dihydrate, sulfuric acid-7h-purin-6-amine-water 1/2/2
bis(7H-purin-6-amine) sulfuric acid dihydrate
Safety and Handling

Safety and Handling

EINECSNumber : 206-286-5

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions