missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Adenine Hydrochloride, ∼99%, MP Biomedicals™
Adenine Hydrochloride, ∼99% is used in viral research and has a wide range of chemical and biochemical roles in vivo and in vitro.
Supplier: MP Biomedicals Inc 0210019180
Description
- Adenine has a wide range of chemical and biochemical roles in vivo and in vitro.
- It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).
- It is used as a blood preservative and in viral research.
- It is a purine nucleobase widespread throughout animal and plant tissues combined with niacinamide, D-ribose, and phosphoric acids.
- It is a constituent of nucleic acids and coenzymes, such as codehydrase I and II, adenylic acid, coalaninedehydrase.
Spécifications
Adenine hydrochloride | |
λ max 262 nm (0.1 N HCl) | |
0.992 | |
MFCD11046306 | |
7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride | |
O.Cl.NC1=C2NC=NC2=NC=N1 | |
189.60 | |
171.6 | |
Powder |
6055-72-7 | |
White | |
C5H8ClN5O | |
100 g | |
MYRDTAUFFBYTHA-UHFFFAOYSA-N | |
7H-purin-6-amine hydrate hydrochloride | |
24211900 | |
99% | |
Used as a blood preservative and in viral research, Regulatory molecule |
Identifiants chimiques
6055-72-7 | |
189.60 | |
MYRDTAUFFBYTHA-UHFFFAOYSA-N | |
24211900 | |
O.Cl.NC1=C2NC=NC2=NC=N1 |
C5H8ClN5O | |
MFCD11046306 | |
7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride | |
7H-purin-6-amine hydrate hydrochloride |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.