missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Adenine sulfate, 98%, synthetic

Supplier:  Thermo Scientific Chemicals 163630250

 View more versions of this product

Catalog No. AC163630250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Adenine sulfate
321-30-2
0.74 to 0.78 (A250/A260), 0.34 to 0.38 (A280/A260)
285.0°C
2% max. (1 g, 105°C, 5 hrs) (vacuum)
98%
C10H12N10O4S
25 g
adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
97.5% min. (lambda max. 262nm,pH 1) molar absorptivity 13100+/-2%,on dry basis
OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
95% min. (c=0.5, H2O, 430nm) 1cm cell
9449
98%
98%
97.5
100.0
White to Yellow
Authentic
Plastic bottle
MFCD00213655
26, V,16, 100
Solubility in water: soluble
LQXHSCOPYJCOMD-UHFFFAOYSA-N
7H-purin-6-amine;sulfuric acid
368.33
184.17
Powder

Chemical Identifiers

321-30-2
368.33
LQXHSCOPYJCOMD-UHFFFAOYSA-N
9449
OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
C10H12N10O4S
MFCD00213655
adenine sulfate, adenine hemisulfate, 7h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
7H-purin-6-amine;sulfuric acid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 206-286-5

RTECSNumber : AU7140000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only