missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Adenosine-2',3'-cyclic monophosphate sodium salt, 97%, Thermo Scientific Chemicals
$407.23 - $920.40
Chemical Identifiers
CAS | 37063-35-7 |
---|---|
Molecular Formula | C10H11N5NaO6P |
Molecular Weight (g/mol) | 351.19 |
MDL Number | MFCD00005757 |
InChI Key | VSDSIACSNXHGOV-GWKNMROSNA-M |
Synonym | adenosine-2 inverted exclamation mark ,3 inverted exclamation mark-cyclic monophosphate sodium salt |
PubChem CID | 131698406 |
IUPAC Name | [(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;sodium |
SMILES | [Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ60558MC
|
Thermo Scientific Chemicals
J60558MC |
100 mg |
Each for $407.23
|
|
||||||
AAJ60558MD
|
Thermo Scientific Chemicals
J60558MD |
250 mg |
Each for $920.40
|
|
||||||
Description
Adenosine-2â,3â-cyclic Monophosphate is an Adenosine (A280400) derivative useful as anticancer agent. It is also used as a Pharmaceutical intermediate, pharmaceutical research and chemical analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdenosine-2â,3â-cyclic Monophosphate is an Adenosine (A280400) derivative useful as anticancer agent. It is also used as a Pharmaceutical intermediate, pharmaceutical research and chemical analysis.
Solubility
It is soluble in DMSO, water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away fom strong oxidizing agents.
Chemical Identifiers
37063-35-7 | |
351.19 | |
VSDSIACSNXHGOV-GWKNMROSNA-M | |
131698406 | |
[Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1 |
C10H11N5NaO6P | |
MFCD00005757 | |
adenosine-2 inverted exclamation mark ,3 inverted exclamation mark-cyclic monophosphate sodium salt | |
[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;sodium |
Specifications
37063-35-7 | |
White | |
MFCD00005757 | |
adenosine-2 inverted exclamation mark ,3 inverted exclamation mark-cyclic monophosphate sodium salt | |
VSDSIACSNXHGOV-GWKNMROSNA-M | |
[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;sodium | |
131698406 | |
97% | |
Adenosine-2',3'-cyclic monophosphate sodium salt |
241°C to 243°C | |
C10H11N5NaO6P | |
100 mg | |
It is soluble in DMSO,water. | |
[Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1 | |
351.19 | |
351.19 | |
Powder |
Safety and Handling
EINECSNumber : 253-328-3
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only