missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adenosine-5'-diphosphate disodium salt, 96% (dry wt.), water 15% max.
CAS: 16178-48-6 | C10H13N5Na2O10P2 | 471.17 g/mol
$64.96 - $143.52
Chemical Identifiers
| CAS | 16178-48-6 |
|---|---|
| Molecular Formula | C10H13N5Na2O10P2 |
| Molecular Weight (g/mol) | 471.17 |
| MDL Number | MFCD00150927 |
| InChI Key | ORKSTPSQHZNDSC-WCYUCLFNNA-L |
| Synonym | adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate |
| PubChem CID | 85315 |
| IUPAC Name | disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate |
| SMILES | [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL14029MD
|
Thermo Scientific Chemicals
L14029MD |
250 mg |
Each for $64.96
|
|
|||||
|
AAL1402903
|
Thermo Scientific Chemicals
L1402903 |
1 g |
Each for $143.52
|
|
|||||
Description
Adenosine-5'-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdenosine-5′-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is -20°C. Store under inert gas. Moisture sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 16178-48-6 | |
| 471.17 | |
| ORKSTPSQHZNDSC-WCYUCLFNNA-L | |
| 85315 | |
| [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
| C10H13N5Na2O10P2 | |
| MFCD00150927 | |
| adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate | |
| disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate |
Specifications
| 16178-48-6 | |
| MFCD00150927 | |
| 3645656 | |
| adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate | |
| ORKSTPSQHZNDSC-WCYUCLFNNA-L | |
| [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
| 471.17 | |
| 471.17 | |
| Adenosine-5'-diphosphate disodium salt |
| C10H13N5Na2O10P2 | |
| 250 mg | |
| Hygroscopic | |
| Soluble in water. | |
| −30° (c=0.5 in 0.5M Na2HPO4) | |
| disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate | |
| 85315 | |
| 97% | |
| (dry wt.), water ≤7% |
Safety and Handling
EINECSNumber : 240-314-7
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only