missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Adenosine 5'-Diphosphate Disodium Salt Hydrate 98.0+%, TCI America™
$51.90 - $152.25
Chemical Identifiers
CAS | 16178-48-6 |
---|---|
Molecular Formula | C10H13N5Na2O10P2 |
Molecular Weight (g/mol) | 471.17 |
MDL Number | MFCD00150927 |
InChI Key | ORKSTPSQHZNDSC-WCYUCLFNNA-L |
Synonym | adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate |
PubChem CID | 85315 |
IUPAC Name | disodium hydrogen ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
SMILES | [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A0626100MG
|
TCI America
A0626100MG |
100 mg |
Each for $51.90
|
|
|||||
A06261G
|
TCI America
A06261G |
1 g |
Each for $152.25
|
|
|||||
Chemical Identifiers
16178-48-6 | |
471.17 | |
ORKSTPSQHZNDSC-WCYUCLFNNA-L | |
85315 | |
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
C10H13N5Na2O10P2 | |
MFCD00150927 | |
adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate | |
disodium hydrogen ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
Specifications
16178-48-6 | |
C10H13N5Na2O10P2 | |
100 mg | |
ORKSTPSQHZNDSC-WCYUCLFNNA-L | |
disodium hydrogen ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate | |
85315 | |
≥98.0% (N) | |
Adenosine 5′-Diphosphate Disodium Salt Hydrate |
White-Yellow | |
MFCD00150927 | |
adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate | |
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
471.17 | |
471.17 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0047
TSCA : Yes
Recommended Storage : Freezer