missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine 5'-monophosphate, 99%
CAS: 61-19-8 | C10H14N5O7P | 347.22 g/mol
$106.68 - $933.61
Identifiants chimiques
| CAS | 61-19-8 |
|---|---|
| Molecular Formula | C10H14N5O7P |
| Molecular Weight (g/mol) | 347.22 |
| MDL Number | MFCD00005750 |
| InChI Key | UDMBCSSLTHHNCD-YPLCUDRINA-N |
| Synonym | adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside |
| PubChem CID | 6083 |
| ChEBI | CHEBI:16027 |
| IUPAC Name | [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
| SMILES | NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC102790050
|
Thermo Scientific Chemicals
102790050 |
5 g | Glass bottle |
chaque for $106.68
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC102790250
|
Thermo Scientific Chemicals
102790250 |
25 g | Glass bottle |
chaque for $331.75
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC102791000
|
Thermo Scientific Chemicals
102791000 |
100 g | Glass bottle |
chaque for $933.61
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 61-19-8 | |
| 347.22 | |
| UDMBCSSLTHHNCD-YPLCUDRINA-N | |
| 6083 | |
| [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
| C10H14N5O7P | |
| MFCD00005750 | |
| adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside | |
| CHEBI:16027 | |
| NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
Spécifications
| 61-19-8 | |
| 0.76 to 0.80 (A250/A260), 0.14 to 0.16 (A280/A260) | |
| 178.0°C to 185.0°C | |
| 6% max. (105°C, 5 hrs) | |
| 98.5% min. (HPLC) | |
| C10H14N5O7P | |
| 5 g | |
| 15, 146 | |
| Solubility in water: soluble. Other solubilities: readily soluble in boiling water, soluble in ethanol | |
| UDMBCSSLTHHNCD-YPLCUDRINA-N | |
| [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate | |
| 347.22 | |
| CHEBI:16027 | |
| 99% | |
| Adenosine 5′-monophosphate |
| 98.5 | |
| 100.0 | |
| Colorless to White | |
| Authentic | |
| Glass bottle | |
| MFCD00005750 | |
| 26, IV, 3615 | |
| adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside | |
| 98% min. (lambda max. 260nm,pH 7) molar absorptivity 15400+/-2%,on dry basis | |
| NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
| 95.0% min. (c=1, 0.2N NaOH, 430nm) 1cm cell | |
| 6083 | |
| 347.21 | |
| Crystalline Powder |
Sécurité et manipulation
missing translation for 'einecsNumber' : 200-500-
RUO – Research Use Only