Learn More
Adenosine-5'-monophosphate disodium salt, Thermo Scientific Chemicals
$59.42 - $165.84
Chemical Identifiers
CAS | 4578-31-8 |
---|---|
Molecular Formula | C10H12N5Na2O7P |
Molecular Weight (g/mol) | 391.19 |
MDL Number | MFCD00065023 |
InChI Key | QGXLVXZRPRRCRP-IDIVVRGQSA-L |
Synonym | adenosine 5'-monophosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate, disodium amp, amp disodium salt, disodium 5'-amp, adenosine phosphate disodium, 5'-amp disodium salt, 5'-adenylic acid, disodium salt, unii-t1wz11dsrn, adenylic acid disodium salt |
PubChem CID | 20712 |
ChEBI | CHEBI:81690 |
IUPAC Name | disodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
SMILES | [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6164306
|
Thermo Scientific Chemicals
J6164306 |
5 g |
Each for $59.42
|
|
|||||
AAJ6164314
|
Thermo Scientific Chemicals
J6164314 |
25 g |
Each for $165.84
|
|
|||||
Description
Adenosine-5'-monophosphate disodium salt is a nucleotide involved in the reactions of cellular energy transfers. It is useful for skin rejuvenation in dermatology and aesthetic dermatology. It acts as an activator of a class of protein kinases called as AMP-activated protein kinase (AMPK). Further, it is used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5'-nucleotidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdenosine-5′-monophosphate disodium salt is a nucleotide involved in the reactions of cellular energy transfers. It is useful for skin rejuvenation in dermatology and aesthetic dermatology. It acts as an activator of a class of protein kinases called as AMP-activated protein kinase (AMPK). Further, it is used as a substrate for enzymes such as AMP-thymidine kinase, AMP deaminase and 5′-nucleotidase.
Solubility
Soluble in water, dimethyl sulfoxide and methanol.
Notes
Air and moisture sensitive. Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
4578-31-8 | |
391.19 | |
QGXLVXZRPRRCRP-IDIVVRGQSA-L | |
20712 | |
disodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
C10H12N5Na2O7P | |
MFCD00065023 | |
adenosine 5'-monophosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate, disodium amp, amp disodium salt, disodium 5'-amp, adenosine phosphate disodium, 5'-amp disodium salt, 5'-adenylic acid, disodium salt, unii-t1wz11dsrn, adenylic acid disodium salt | |
CHEBI:81690 | |
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
Specifications
4578-31-8 | |
C10H12N5Na2O7P | |
5 g | |
14,158 | |
Soluble in water,dimethyl sulfoxide and methanol. | |
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
391.19 | |
CHEBI:81690 | |
Powder |
White | |
MFCD00065023 | |
3586732 | |
adenosine 5'-monophosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate, disodium amp, amp disodium salt, disodium 5'-amp, adenosine phosphate disodium, 5'-amp disodium salt, 5'-adenylic acid, disodium salt, unii-t1wz11dsrn, adenylic acid disodium salt | |
QGXLVXZRPRRCRP-IDIVVRGQSA-L | |
disodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate | |
20712 | |
391.2 | |
Adenosine-5'-monophosphate disodium salt |
Safety and Handling
EINECSNumber : 224-961-2
RTECSNumber : AU7481600
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only