missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adenosine-5'-Triphosphate Disodium Salt Hydrate MP Biomedicals

Adenosine-5'-triphosphate Disodium Salt Hydrate Cell culture reagent, ≥96%, White Powder

Supplier:  MP Biomedicals Inc 0215026680

 View more versions of this product

Catalog No. ICN15026680


Explore available promotions

Description

Description

  • Adenosine 5'-triphosphate (ATP) and its phosphate bonds are the basic components of energy exchange in many biological systems.
Specifications

Specifications

Adenosine-5'-Triphosphate Disodium Salt Hydrate
White
Powder
C10H20N5Na2O16P3
adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt
O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
605.19
551.2 (Anhydrous)
Hydrate
51963-61-2
100 g
MFCD00150754
MWEQTWJABOLLOS-MVHBFMHHNA-L
disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
134129625

Chemical Identifiers

51963-61-2
605.19
MWEQTWJABOLLOS-MVHBFMHHNA-L
O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
C10H20N5Na2O16P3
MFCD00150754
134129625
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 0°C.

SDS
Documents

Documents

Promotions

Promotions