missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adenosine, 99+%
CAS: 58-61-7 | C10H13N5O4 | 267.25 g/mol
$78.11 - $182.32
Chemical Identifiers
| CAS | 58-61-7 |
|---|---|
| Molecular Formula | C10H13N5O4 |
| Molecular Weight (g/mol) | 267.25 |
| MDL Number | MFCD00005752 |
| InChI Key | OIRDTQYFTABQOQ-KQYNXXCUSA-N |
| Synonym | adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol |
| PubChem CID | 60961 |
| ChEBI | CHEBI:16335 |
| IUPAC Name | (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| SMILES | NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC164040050
|
Thermo Scientific Chemicals
164040050 |
5 g | Glass bottle |
N/A
|
|
||||
|
AC164040250
|
Thermo Scientific Chemicals
164040250 |
25 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 58-61-7 | |
| 267.25 | |
| OIRDTQYFTABQOQ-KQYNXXCUSA-N | |
| 60961 | |
| (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| C10H13N5O4 | |
| MFCD00005752 | |
| adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol | |
| CHEBI:16335 | |
| NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1 |
Specifications
| 58-61-7 | |
| 233.0°C to 238.0°C | |
| 0.5% (105°C, 2 hrs) max. | |
| 99% min. (HPLC) | |
| C10H13N5O4 | |
| 5 g | |
| 15141 | |
| adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol | |
| 98.0 to 102% (lambda max. 259nm,pH 7) molar absorptivity 15400+/-2% ,on dry basis | |
| NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1 | |
| 98% (c=0.5, H2O, 430nm) 1cm cell min. | |
| 60961 | |
| 267.23 | |
| Crystalline Powder |
| 0.76 to 0.80 (A250/A260),0.14 to 0.16 (A280/A260) | |
| White | |
| Authentic | |
| Glass bottle | |
| MFCD00005752 | |
| 26, V,16, 335 | |
| − 60.00 | |
| Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
| OIRDTQYFTABQOQ-KQYNXXCUSA-N | |
| (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol | |
| 267.25 | |
| CHEBI:16335 | |
| 99+% | |
| Adenosine |
Safety and Handling
EINECSNumber : 200-389-9
RUO – Research Use Only