missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Adenosine, 99.8%, MP Biomedicals™
A nucleotide resulting from the union of one molecule of sugar -- d-ribose -- with a base, l-adenine
$58.69 - $2976.49
Chemical Identifiers
CAS | 58-61-7 |
---|---|
Molecular Formula | C10H13N5O4 |
Molecular Weight (g/mol) | 267.25 |
MDL Number | MFCD00005752 |
InChI Key | OIRDTQYFTABQOQ-KQYNXXCUSA-N |
Synonym | adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol |
PubChem CID | 60961 |
ChEBI | CHEBI:16335 |
SMILES | NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10019905
|
MP Biomedicals Inc
0210019905 |
5 g |
Each for $58.69
|
|
|||||
ICN10019991
|
MP Biomedicals Inc
0210019991 |
1 kg |
Each for $2,976.49
|
|
|||||
Chemical Identifiers
58-61-7 | |
267.25 | |
OIRDTQYFTABQOQ-KQYNXXCUSA-N | |
60961 | |
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1 |
C10H13N5O4 | |
MFCD00005752 | |
adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol | |
CHEBI:16335 |
Specifications
58-61-7 | |
2.09g/cm3 | |
C10H13N5O4 | |
5 g | |
OIRDTQYFTABQOQ-KQYNXXCUSA-N | |
(2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol | |
60961 | |
267.2 | |
Adenosine |
White | |
99.80% | |
MFCD00005752 | |
adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol | |
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1 | |
267.25 | |
CHEBI:16335 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : 200-389-9