missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Adipic acid monoethyl ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2400614
Description
Adipic acid monoethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdipic acid monoethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Adipic acid monoethyl ester | |
28°C to 29°C | |
179°C to 181°C (decomposition) | |
C8H14O4 | |
MFCD00004419 | |
adipic acid monoethyl ester, monoethyl adipate, ethyl hydrogen adipate, hexanedioic acid, monoethyl ester, monoethyl hexanedioate, monoethyladipic acid ester, adipic acid, monoethyl ester, mono-ethyl adipate, hexanedioic acid, 1-ethyl ester, hexanoic acid, monoethyl ester | |
UZNLHJCCGYKCIL-UHFFFAOYSA-N | |
6-ethoxy-6-oxohexanoic acid | |
12295 | |
95% |
626-86-8 | |
0.98 | |
>110°C (230°F) | |
1.439 | |
25 g | |
Soluble in water. | |
CCOC(=O)CCCCC(=O)O | |
174.196 | |
174.2 |
Chemical Identifiers
626-86-8 | |
174.196 | |
UZNLHJCCGYKCIL-UHFFFAOYSA-N | |
12295 | |
CCOC(=O)CCCCC(=O)O |
C8H14O4 | |
MFCD00004419 | |
adipic acid monoethyl ester, monoethyl adipate, ethyl hydrogen adipate, hexanedioic acid, monoethyl ester, monoethyl hexanedioate, monoethyladipic acid ester, adipic acid, monoethyl ester, mono-ethyl adipate, hexanedioic acid, 1-ethyl ester, hexanoic acid, monoethyl ester | |
6-ethoxy-6-oxohexanoic acid |
Safety and Handling
EINECSNumber : 210-966-7
RTECSNumber : AV2030000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only