missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adipic dihydrazide, 98%, Thermo Scientific Chemicals

$131.05 - $487.51

Chemical Identifiers

CAS 1071-93-8
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00007614
InChI Key IBVAQQYNSHJXBV-UHFFFAOYSA-N
Synonym adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
PubChem CID 66117
IUPAC Name hexanedihydrazide
SMILES NNC(=O)CCCCC(=O)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC203310250
SDS View Documents
Thermo Scientific Chemicals
203310250
25 g Plastic bottle
Each for $131.05
 
AC203311000
SDS View Documents
Thermo Scientific Chemicals
203311000
100 g Plastic bottle
Each for $487.51
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1071-93-8
174.20
IBVAQQYNSHJXBV-UHFFFAOYSA-N
66117
NNC(=O)CCCCC(=O)NN
C6H14N4O2
MFCD00007614
adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
hexanedihydrazide

Specifications

1071-93-8
100.0
White to Yellow
98%
C6H14N4O2
MFCD00007614
02, I, 277
Solubility in water: soluble. Other solubilities: soluble in acetic acid,slightly soluble in acetone,insoluble in ethanol,ether and benzene
NNC(=O)CCCCC(=O)NN
174.20
174.2
Crystalline Powder
97.5
178.0°C to 182.0°C
Authentic
Plastic bottle
H2NNHCO(CH2)4CONHNH2
25 g
adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
IBVAQQYNSHJXBV-UHFFFAOYSA-N
hexanedihydrazide
66117
98%
Adipic dihydrazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 213-999-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only