Adjustable Tuning Fork
Precision-grade aluminum alloy.
Supplier: Customer Channels Group (Ccg) 700240ADJ
Description
- Sliding adjustable weights on each prong
- Marked to deliver tones at C-128, D-144, E-160, F-170.7, G-192, A-213.3 and B-240 frequencies
Specifications
Tuning Fork, Adjustable | |
C-128, D-144, E-160, F-170.7, G-192, A-213.3 and B-240 frequencies |