MilliporeSigma™ Supelco™ Adustable Single Port Mini-Vap Evaporator/Concentrator Mini-Vap
Ideal for a wide variety of needs in evaporation or concentration.
Supplier:  MilliporeSigma™ Supelco™ 22970
Catalog No. 111020810
Explore available promotions
Description
Description
The Mini-Vap includes a needle valve for fine metering of air or nitrogen drying gas. Ideal for a wide variety of needs in evaporation or concentration. The adjustable Mini-Vap (22970) is a versatile concentrator/evaporator that can be used with a single container. The unique design allows for any size container, from the smallest vial to a 250 mL beaker.
Specifications
Specifications
1/Pk.
Size: 1 1/2 in. 8 1/2 in., adjustable, single unit