Advanced Instruments Renol™ Urine Control Solution
Supplier: Advanced Instruments 3LA085
Description
- For use with Advanced Osmometers: 3320, 2020, 3250, 3900, and 210
Specifications
CE, CLIA, CAP | |
300 and 800 mOsm/kg H2O | |
2 Level Urine Control Kit | |
Evaluating the performance the osmometer in clinical laboratories testing Human Urine |
2°C to 8°C | |
2020, 3D3, 3250, 3300 and 3320 Urine Osmometer | |
3mL Vials and 4 vials per level | |
8 Pack |