Learn More
METTLER TOLEDO Advanced MR Analytical Balance
METTLER TOLEDO Advanced MR Analytical Balances offer a highly precise weigh cell with FACT automatic internal adjustment.
$6,480.89 - $7,476.89
Specifications
Connection Type USB-A, USB-C, RS232, Ethernet
Display Type Color Touchscreen
Readability 0.1 mg
Material Metal
Stabilization Time 2 sec.
View More Specs
Products 3
Catalog Number Mfr. No. Capacity Model Price Quantity  
Catalog Number Mfr. No. Capacity Model Price Quantity  
01-804-284
View Documents Promotion Details
METTLER TOLEDO
30666203
120 g MR104 N/A
Only null left
01-804-282
View Documents Promotion Details
METTLER TOLEDO
30666206
220 g MR204 N/A
Only null left
01-804-280
View Documents Promotion Details
METTLER TOLEDO
30666209
320 g MR304 N/A
Only null left
Description
Description
User management within the 4.5-inch touchscreen increases efficiency of daily tasks with 8 built-in applications and statistical data analysis. Designed with full metal housing and overload protection, these balances will provide high quality performance for years to come. When your balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
MR Analytical Balances include:
  • FACT automatic internal adjustment
  • 4.5 inch color touchscreen
  • 8 built-in applications
  • Statistical data analysis
  • User management
  • Guided leveling
  • 4 interfaces: USB-A, USB-C, RS232, Ethernet
  • Bluetooth option
  • Full metal housing
  • Overload protection
The MR Balance is the successor to the ML or ME-T Balance. See how it ranks among our balance lines:
  • XPR Excellence Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • ME Standard Analytical Balances
  • LA Standard Analytical Balances
Specifications
Specifications
USB-A, USB-C, RS232, Ethernet
0.1 mg
2 sec.
100/240 V, 50/60 Hz
Color Touchscreen
Metal
Analytical Balance
No
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'