Learn More
METTLER TOLEDO Advanced MX Precision Balance
METTLER TOLEDO Advanced MX Precision Balances feature a precise weigh cell and automatic internal adjustment. The SmartPan stabilizes in any weighing environment, ensuring you receive consistently accurate results.
$6378.98 - $14861.12
Specifications
Connection Type USB-A, USB-C, Ethernet
Display Type Color Touchscreen
Material Metal
Type Precision Balance
Electrical Requirements 100/240 V, 50/60 Hz
View More Specs
Products 17
Catalog Number Mfr. No. Capacity Readability Model Price Quantity  
Catalog Number Mfr. No. Capacity Readability Model Price Quantity  
01-804-318
View Documents Promotion Details
METTLER TOLEDO
30665149
320 g 1 mg MX303
Each for $7,481.53
Only null left
01-804-316
View Documents Promotion Details
METTLER TOLEDO
30665152
320 g 1 mg MX303N
Each for $6,378.98
Only null left
01-804-314
View Documents Promotion Details
METTLER TOLEDO
30665155
620 g 1 mg MX603
Each for $8,977.82
Only null left
01-804-312
View Documents Promotion Details
METTLER TOLEDO
30665158
620 g 1 mg MX603N
Each for $7,652.93
Only null left
01-804-310
View Documents Promotion Details
METTLER TOLEDO
30665161
1220 g 1 mg MX1203 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-804-308
View Documents Promotion Details
METTLER TOLEDO
30665164
1220 g 1 mg MX1203N
Each for $9,139.97
Only null left
01-804-306
View Documents Promotion Details
METTLER TOLEDO
30665167
2.2 kg 0.01 g MX2002
Each for $7,064.60
Only null left
01-804-304
View Documents Promotion Details
METTLER TOLEDO
30665170
4.2 kg 0.01 g MX4002
Each for $8,366.33
Only null left
01-804-302
View Documents Promotion Details
METTLER TOLEDO
30665173
6.2 kg 0.01 g MX6002
Each for $9,204.81
Only null left
01-804-300
View Documents Promotion Details
METTLER TOLEDO
30665176
1.2 kg, 6.2 kg 0.01 g, 0.1 g MX6002DR
Each for $8,282.94
Only null left
01-804-298
View Documents Promotion Details
METTLER TOLEDO
30665179
12.2 kg 0.01 g MX12002 N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-804-296
View Documents Promotion Details
METTLER TOLEDO
30665182
6.2 kg 0.1 g MX6001
Each for $6,902.46
Only null left
01-804-294
View Documents Promotion Details
METTLER TOLEDO
30665185
8.2 kg 0.1 g MX8001
Each for $7,847.50
Only null left
01-804-292
View Documents Promotion Details
METTLER TOLEDO
30665188
12.2 kg 0.1 g MX12001L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-804-290
View Documents Promotion Details
METTLER TOLEDO
30665191
16.2 kg 0.1 g MX16001L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-804-288
View Documents Promotion Details
METTLER TOLEDO
30665194
32.2 kg 0.1 g MX32001L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-804-286
View Documents Promotion Details
METTLER TOLEDO
30665197
32.2 kg 1 g MX32000L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Exceptional user management within the 7-inch touchscreen allows easy access to 9 built-in weighing applications, assigns user access rights and provides automatic data logging. These balances are designed with full metal housing and IP54 protection to defend against environmental influences and rough handling. The power-saving mode can be enabled when the balance is not in use to save costs and reduce energy usage.
MX Precision Balances include:
  • FACT automatic internal adjustment
  • SmartPan
  • 7-inch touchscreen
  • 9 built-in applications
  • Statistical data analysis
  • User management
  • Leveling assistant
  • StatusLight
  • 3 interfaces: USB-A, USB-C, Ethernet
  • Bluetooth option
  • EasyDirect software
  • Full metal housing with overload protection
The MX Balance is the successor to the MS or XSR Balance. See how it ranks among our balance lines:
  • XPR Excellence Precision Balances
  • MX Advanced Precision Balances
  • MR Advanced Precision Balances
  • MA Standard Precision Balances
  • ME Standard Precision Balances
  • LA Standard Precision Balances
Specifications
Specifications
USB-A, USB-C, Ethernet
Metal
100/240 V, 50/60 Hz
Color Touchscreen
Precision Balance
No
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'