Learn More
ZOLL™ Medical AED Plus™ Semiautomatic Defibrillator Packages
Provides real CPR help for rate and depth of chest compressions
$4431.50 - $4431.50
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
17-300-100
|
ZOLL™ Medical
22600010102011060 |
ZOLL AED Plus w/AED cover, PlusRX medical prescription, CPR-D-padz electrode, pack of 10 batteries, and carry case |
Each for $4,431.50
|
|
|||||
Description
The AED Plus™ is the first and only full-rescue public access AED that provides Real CPR Help™ for rate and depth of chest compressions. Real-time feedback helps resucers provide the best manual CPR possible in a sudden cardiac arrest (SCA). The AED Plus also guides rescuers through the complete Chain of Survival, helping all SCA victims, not just those who need a shock. The AED Plus features ZOLL's unique one-piece electrode pad for fast and accurate placement and is powered by lithium camera batteries available from retail stores. Both the pad and the batteries have a five-year shelf life.
- Comes with Medical Prescription or PlusTrac Professional1 or PlusTrac Professional5
AED Plus w/Medical Prescription (Model 8000-004000-01) includes:
- ZOLL AED Plus w/AED Cover, PlusRX Medical Prescription, CPR-D padz™ Electrode, pack of 10 CR123a batteries, and Carry Case
- ZOLL AED Plus w/AED Cover, PlusTrac Professional1, CPR-D-padz Electrode, pack of 10 CR123a batteries, and Carry Case
- ZOLL AED Plus w/AED Cover, PlusTrac Professional5, CPR-D-padz Electrode, pack of 10 CR123a batteries, and Carry Case
Specifications
Defibrillator Package |