missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aescin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6696809

 View more versions of this product

Catalog No. AAJ6696809


Explore available promotions

Description

Description

It is used and studied as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.1 It may be used to study its pharmacokinetics, safety and efficacy as a chemosensitizer and modulator of NF-κ B cell signaling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used and studied as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.1 It may be used to study its pharmacokinetics, safety and efficacy as a chemosensitizer and modulator of NF-κ B cell signaling

Solubility
Soluble in methanol.

Notes
Desiccate at room temperature. Incompatible with oxidizing agents.
Specifications

Specifications

Aescin
White
MFCD00076054
14,3696
Soluble in DMSO.
CC=C(C)C(=O)OC1C(C2(C(CC1(C)C)C3=CCC4C5(CCC(C(C5CCC4(C3(CC2O)C)C)(C)CO)OC6C(C(C(C(O6)C(=O)O)OC7C(C(C(C(O7)CO)O)O)O)O)OC8C(C(C(C(O8)CO)O)O)O)C)CO)OC(=O)C
1131.269
1131.26
Powder
6805-41-0
C55H86O24
10 g
aescin
AXNVHPCVMSNXNP-XJVYZZIXSA-N
(2S,3S,4S,5R,6R)-6-[[(3S,4S,6aR,6bS,8S,8aR,9R,10R,14bR)-9-acetyloxy-8-hydroxy-4,8a-bis(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-10-[(Z)-2-methylbut-2-enoyl]oxy-1,2,3,4a,5,6,7,8,9,10,12,12a,14,14a-tetradecahydropicen-3-yl]oxy]-4-hydroxy-3-[(2R,3S,4R,5R,
133640184
98%

Chemical Identifiers

6805-41-0
1131.269
AXNVHPCVMSNXNP-XJVYZZIXSA-N
133640184
CC=C(C)C(=O)OC1C(C2(C(CC1(C)C)C3=CCC4C5(CCC(C(C5CCC4(C3(CC2O)C)C)(C)CO)OC6C(C(C(C(O6)C(=O)O)OC7C(C(C(C(O7)CO)O)O)O)O)OC8C(C(C(C(O8)CO)O)O)O)C)CO)OC(=O)C
C55H86O24
MFCD00076054
aescin
(2S,3S,4S,5R,6R)-6-[[(3S,4S,6aR,6bS,8S,8aR,9R,10R,14bR)-9-acetyloxy-8-hydroxy-4,8a-bis(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-10-[(Z)-2-methylbut-2-enoyl]oxy-1,2,3,4a,5,6,7,8,9,10,12,12a,14,14a-tetradecahydropicen-3-yl]oxy]-4-hydroxy-3-[(2R,3S,4R,5R,
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c

H302+H332-H319-H335

EINECSNumber : 229-880-6

RTECSNumber : KF6296000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only