missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Agar, NF Grade
Powder, meets N.F. grade
Supplier: MP Biomedicals™ 0210026283
Description
- 80-100 mesh
- Substitute for gelatin, isinglass
- Used as nutrient media for bacterial culture
Specifications
15°C to 30°C | |
USP, NF | |
Used for nutrient media for bacterial cultures |
Powder | |
Agar | |
250 g |