missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agar, Powder, NF, Spectrum™ Chemical

AG110, 9002-18-0

Supplier:  Spectrum Chemical Mfg Cor AG11025KGBL

 View more versions of this product

Catalog No. 18600338

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Spectrum™ Chemical Agar, Powder, NF is used in the pharmaceutical industry for many applications such as a suspending agent, surgical lubricants and in preparations of emulsions, suspensions and capsules. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

9002-18-0
Fiber Drum
25 kg
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
336.34
100%
C14H24O9
GYYDPBCUIJTIBM-UHFFFAOYNA-N
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
NF

Chemical Identifiers

9002-18-0
336.34
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
C14H24O9
GYYDPBCUIJTIBM-UHFFFAOYNA-N
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
Videos
SDS
Documents

Documents

Promotions

Promotions