missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agarose, Electrophoresis Grade, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210026790

 View more versions of this product

Catalog No. ICN10026790


Explore available promotions

Description

Description

Specifications

Specifications

Agarose
9012-36-6
White
MFCD00081294
Electrophoresis
Powder
1.00g/mL at 20°C
500 g
3, 6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose

Chemical Identifiers

9012-36-6
MFCD00081294
3, 6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature (15 to 30 °C)

SDS
Documents

Documents

Promotions

Promotions