missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
Agarose, C12H18O9, CAS Number-9012-36-6, 500g, Gray, Cream, DNase free, 0.09 to 0.13, Pass Test, 34.5 deg. to 37.5 deg.C, 1200g/cm2min., Molecular Biology, Low-EEO/Multi-Purpose, Poly Bottle, Powder/Solid, DNase- and RNase-Free, Room temperature, 0.15% max.
$197.98 - $918.73
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP160-100
|
Fisher BioReagents
BP160100 |
100 g | Poly Bottle |
Each for $197.98
Case of 6 Each for $1,187.88
|
|
||||
BP160-500
|
Fisher BioReagents
BP160500 |
500 g | Poly Bottle |
Each for $918.73
Case of 6 Each for $5,512.38
|
|
||||
Description
Agarose, C12H18O9, CAS Number-9012-36-6, 500g, Gray, Cream, DNase free, 0.09 to 0.13, Pass Test, 34.5 deg. to 37.5 deg.C, 1200g/cm2min., Molecular Biology, Low-EEO/Multi-Purpose, Poly Bottle, Powder/Solid, DNase- and RNase-Free, Room temperature, 0.15% max.
Chemical Identifiers
9012-36-6 | |
C12H18O9 |
Specifications
Powder/Solid | |
Cream | |
DNase free | |
34.5° to 37.5°C | |
Poly Bottle | |
DNase- and RNase-Free |
9012-36-6 | |
100 g | |
0.09 to 0.13 | |
1200g/cm2min. | |
C12H18O9 | |
Agarose, Low-EEO/Multi-Purpose |
Safety and Handling
Recommended Storage : Room temperature