missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Agarose (Low-Melting, Nucleic Acid Recovery/Molecular Biology Grade), Fisher BioReagents
Supplier: Fisher BioReagents BP16525
Specifications
Agarose | |
Powder/Solid | |
Cream | |
DNase- and RNase-Free | |
≤0.10 | |
26° to 30°C | |
306.12 | |
Poly Bottle |
Low-Melting, Nucleic Acid Recovery | |
39346-81-1 | |
25 g | |
DNase free | |
C12H18O9 | |
≥200g/cm2 | |
Molecular Biology |
Chemical Identifiers
39346-81-1 | |
C12H18O9 |
Safety and Handling
Recommended Storage : RT