Walter Stern Agate Mortar and Pestle Sets
Ideal for preparing laboratory samples and high purity powders.
$278.31 - $4033.94
Specifications
Finish Highly Polished Grinding Surface
Product Type Mortar and Pestle
Material Agate Stone
For Use With (Application) For grinding or crushing
View More Specs
Products 7
Catalog Number Mfr. No. Diameter (English) Outer Diameter (Metric) Outer Height (English) Height (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Outer Diameter (Metric) Outer Height (English) Height (Metric) Price Quantity  
12-950AA
View Documents
Walter Stern
400D
2 in. 5.08cm 0.9 in. 2.3cm
Each for $278.31
12-950A
View Documents
Walter Stern
400E
2.55 in. 6.5cm 1.02 in. 2.6cm
Each for $383.44
12-950BB
View Documents
Walter Stern
400F
3 in. 7.62cm 1.18 in. 3cm
Each for $538.69
12-950B
View Documents
Walter Stern
400G
3.75 in. 9.52cm 1.57 in. 4cm
Each for $873.50
12-950C
View Documents
Walter Stern
400H
4 in. 10.16cm 1.6 in. 4.1cm
Each for $968.16
12-950D
View Documents
Walter Stern
400J
5 in. 12.7cm 2 in. 5.1cm
Each for $1,926.25
12-950E
View Documents
Walter Stern
400K
6 in. 15.24cm 2.55 in. 6.5cm
Each for $4,033.94
Description
Description
Ideal for preparing laboratory samples and high purity powders.
  • Octagonally shaped, thick walled, agate mortar with pestle
  • Both have highly polished grinding surface
Specifications
Specifications
Highly Polished Grinding Surface
Mortar and Pestle
Agate Stone
For grinding or crushing
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'