BTX™ AgilePulse™ MAX Safety Stand for Flatpack
AgilePulse™ MAX Safety Stand for Flatpack for use with BTX™ AgilePulse™ MAX.
Supplier:  BTX™ 470209
Catalog No. BTX470209
Explore available promotions
Description
Description
  • Cuvette module plugs directly into generator
  • Holds one cuvette
  • Safety swing shield secures cuvette in holder
Specifications
Specifications
Electroporation
Safety Stands
AgilePulse MAX Safety Stand for Flatpack
Safety Stand Only
BNC Cable
AC Sine Wave, DC Square Wave
AgilePulse™ MAX, ECM™ 2001+
500 to 1000 V
Touchscreen
Non-sterile
Digital User