Learn More
Applied Biosystems™ AgPath-ID™ One-Step RT-PCR Reagents
AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.
Supplier: Applied Biosystems™ 4387424
Description
AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.
- Recommended for RNA pathogen amplification
- Consistent amplification of RNA targets with high specificity and sensitivity
- Optimized to work with your target-specific primers and probes
- Contains ROX for quantitative fluorescent signal normalization
AgPath-ID™ One-Step RT-PCR Reagents are configured for fast and simple reaction setup. The reactions are assembled in a single tube, minimizing sample handling errors and expediting set-up time. Once assembled, results are available in approximately one hour. The 25X RT-PCR Enzyme Mix included in the kit contains highly efficient ArrayScript™ Reverse Transcriptase, a mutant MMLV RT that produces high cDNA yields, and AmpliTaq Gold™ polymerase, the preferred hot-start DNA polymerase for specific target amplification. The 2X RT-PCR Buffer has been optimized for efficient, robust reverse transcription and PCR includes the passive reference dye, ROX™ Dye, for quantitative fluorescent signal normalization.
Specifications
Sufficient for 500 reactions of 25 μL, contains: • 7mL 2X RT-PCR Buffer • 550 μL 25X RT-PCR Enzyme Mix • 25 mL Nuclease-free Water. |
|
Primer-probe | |
High | |
AmpliTaq Gold DNA Polymerase | |
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex | |
AgPath-ID™, Ambion™ | |
500 reactions | |
Dry Ice | |
25X | |
500 Reactions | |
7 mL |
Works on all AB platforms, but is optimized for the 7500. | |
Tube | |
1-step RT-qPCR | |
Standard | |
ROX (Pre-mixed) | |
One-Step RT-PCR Reagent | |
ArrayScript™ | |
500 Reactions | |
25 X | |
RNA |
For Research Use Only.