missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ AgPath-ID™ One-Step RT-PCR Reagents DFS Item

AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.

Supplier:  Applied Biosystems™ AM1005

 View more versions of this product

Catalog No. AM1005


Explore available promotions

Description

Description

AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.

  • Recommended for RNA pathogen amplification
  • Consistent amplification of RNA targets with high specificity and sensitivity
  • Optimized to work with your target-specific primers and probes
  • Contains ROX for quantitative fluorescent signal normalization

AgPath-ID™ One-Step RT-PCR Reagents are configured for fast and simple reaction setup. The reactions are assembled in a single tube, minimizing sample handling errors and expediting set-up time. Once assembled, results are available in approximately one hour. The 25X RT-PCR Enzyme Mix included in the kit contains highly efficient ArrayScript™ Reverse Transcriptase, a mutant MMLV RT that produces high cDNA yields, and AmpliTaq Gold™ polymerase, the preferred hot-start DNA polymerase for specific target amplification. The 2X RT-PCR Buffer has been optimized for efficient, robust reverse transcription and PCR includes the passive reference dye, ROX™ Dye, for quantitative fluorescent signal normalization.

Specifications

Specifications

Sufficient for 100 reactions of 25 μL, contains:
• 1375μL 2X RT-PCR Buffer
• 110μL 25X RT-PCR Enzyme Mix
• 1.75mL Nuclease-free Water.
Primer-probe
High
AmpliTaq Gold DNA Polymerase
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
AgPath-ID™, Ambion™
100 reactions
Dry Ice
25X
100 Reactions
1.75 mL
Works on all AB platforms, but is optimized for the 7500.
Tube
1-step RT-qPCR
Standard
ROX (Pre-mixed)
One-Step RT-PCR Reagent
ArrayScript™
100 Reactions
25 X
RNA
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.