Ahlstrom-Munksjo Pleated (Fluted) Filter Paper, Grade 509
Ahlstrom offers a full range of standard pleated grades that meet your needs from grades with rapid flow rates for filtration of viscous materials to grades with tight matrices for removal of fine precipitates.
Supplier:  Ahlstrom-Munksjo 50903200
 View more versions of this product
Catalog No. 36-101-3276
Description
Description
Specifications
Specifications
4 μm
509
32 cm