missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ AIM V™ Medium, liquid (research grade)
AIM V™ Medium, liquid (research grade)
$149.00 - $264.00
Specifications
Content And Storage | Store in refrigerator (2–8°C) and protect from light. |
---|---|
Cell Type | Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes |
Culture Type | Mammalian Cell Culture |
Form | Liquid |
Product Type | Serum Free Medium (SFM) |
Description
Contains L-glutamine, streptomycin sulfate at 50μg/mL, and gentamicin sulfate at 10μg/mL.Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Mammalian Cell Culture | |
Serum Free Medium (SFM) | |
Streptomycin Sulfate, Gentamicin | |
Research Grade |
Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes | |
Liquid | |
Glutamine, Phenol Red, HSA | |
AIM V™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.