missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Air Systems™ Confined Space Ventilation Kit: Accessories
Accessories for Air Systems Confined Space Ventilation Kit
Supplier: Air Systems™ SV90
Description
For use with Air Systems Confined Space Ventilation Kit: (Cat. No. 18-999-331)
Specifications
Orange | |
90° Elbow | |
Saddle Vent™ | |
110.49 cm | |
36.83 cm |
8 in. | |
43.5 x 14.5 in. (110.49 x 36.83 cm) | |
43.5 in. | |
Polyethylene | |
14.5 in. |