missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Air Systems™ Confined Space Ventilation Kit: Accessories DFS Item

Accessories for Air Systems Confined Space Ventilation Kit

Supplier:  Air Systems™ SVH88C

 View more versions of this product

Catalog No. 18-500


Explore available promotions

Description

Description

For use with Air Systems Confined Space Ventilation Kit: (Cat. No. 18-999-331)

Specifications

Specifications

Silver
20.32 cm
Saddle Vent™
8 in.
Duct to Duct Connector, 8 in.
Polyethylene
Videos
SDS
Documents

Documents

Promotions

Promotions