AirClean™ Systems Combination PCR Workstations
The Airclean Combination PCR Workstation combines ISO 5 (FS209E) Class 100 clean air environment with UV light sterilization for optimal protection from sample contamination.
Supplier:  AirClean™ Systems AC632TLFUVC
 View more versions of this product
Catalog No. 36-100-4288
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • UVTect™ microprocessor controller
  • Class 100 (ISO 5) vertical laminar flow air
  • Polycarbonate and polypropylene design to reflect UV energy
  • Full access folding sash
  • Unibody plastic design - no joints or gaps in construction
  • Digital UV light timer 0-59 minutes
  • Lab event timer
  • Available in 24 in. , 32 in., and 48 in. widths; nominal depth 24 in.
  • 110V AC or 220V AC versions (24 in. only offered as 110V)
UVTect™ Features:
  • Variable speed blower control
  • 254nm ultraviolet light with user set digital timer
  • Constant HEPA filter monitoring
  • Constantly monitors and alerts effectiveness of the shortwave UV bulb life
  • Lab event timer
Specifications
Specifications
Combination PCR Workstation
Digital UV
23 x 31 x 27 in. (58.4 x 78.7 x 68.5 cm)