missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alconox™ Tergazyme™ Enzyme-Active Powered Detergent

Concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning

$207.83 - $10211.40

Specifications

Product Type Enzyme-Active Powered Detergent
Form Powder
pH 9.5
Appearance White, Cream
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
View More Specs

Products 5
Catalog Number Mfr. No. Container Type Volume (English) Price Quantity  
Catalog Number Mfr. No. Container Type Volume (English) Price Quantity  
16-000-118
View Documents
Alconox™
ALC1301
Drum 100 lb.
Each for $3,876.87
 
16-000-115
View Documents
Alconox™
1304
Box 4 lb.
Each for $207.83
Case of 9 Each for $1,870.47
 
04-322-11C
View Documents
Alconox™
ALC1350
Carton 50 lb.
Each for $2,018.17
 
16-000-116
View Documents
Alconox™
ALC1325
Carton 25 lb.
Each for $1,071.63
 
16-000-119
View Documents
Alconox™
ALC1303
Drum 300 lb. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Tergazyme consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, phosphates, carbonates, and protease enzyme.

  • Concentrated, biodegradable and disposable
  • Ideal for removal of proteinaceous soils, tissue, blood and body fluids from glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass
  • Replaces corrosive acids and hazardous solvents
  • Free rinsing gives reliable results without interfering residues
  • Dilute 1:100 for typical use (10g in 1L of water)
  • Phosphate Content: 7.5% (as Phosphorus)
  • Use as cleaning agent in Reverse Osmosis and Ultra-Filtration Systems
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • USDA authorized for use in federally inspected meat and poultry plants

Used to clean:
Medical instruments/devices, dairy equipment, laboratory ware, reverse osmosis and ultrafiltration membranes and units, sampling apparatus, pharmaceutical apparatus, cosmetics manufacturing equipment, tubing, pipes, optical parts, process equipment, industrial parts, desalination plants, tanks and reactors. Passes inhibitory residue test for water analysis.

Used to remove:
Soil, grit, grime, blood, tissue, grease, fats, oils, proteinaceous soils, dairy proteins, particulates, solvents and bioreactor residue.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.

Shelf life: 2 years from date of manufacture.

Order Info

0.5kg (1 lb.) makes 49.2L (13 gal.) cleaning solution.

Specifications

Specifications

Enzyme-Active Powered Detergent
9.5
Store closed and upright in a cool dry place at 15° to 30°C.
Powder
White, Cream
Remove proteinaceous soils, tissue, blood and body fluids
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'