Alcor™ Seditrol™ ESR Control Kit
For use with iSED automated ESR analyzer
Supplier: Alcor™ DSC06
Description
- Barcoded
- Bi-level human-based controls
- Remains on-board and is ready for testing 24Ø7
- User selectable auto-QC parameters
- Fulfills unique requirements of laboratory
- Includes 6 x 4.5mL Tubes
Specifications
3 month supply of three Level 1 and three Level 2 samples in 13 x 75mm tubes | |
SEDITROL Quality Control | |
60 day open vial stability |