missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alendronate Sodium Trihydrate 97.0+%, TCI America™

Supplier:  TCI America A24565G

 View more versions of this product

Catalog No. A24565G


Explore available promotions

Description

Description

Specifications

Specifications

Alendronate Sodium Trihydrate
White
MFCD01748233
alendronate sodium hydrate, alendronate sodium trihydrate, alendronate sodium, alendronate monosodium trihydrate, alendros, adronat, fosamax, sodium alendronate hydrate, elandor, fosalan
C(CC(O)(P(=O)(O)O)P(=O)(O)[O-])CN.O.O.O.[Na+]
325.122
CHEBI:2566
≥97.0% (T)
121268-17-5
C4H18NNaO10P2
5 g
DCSBSVSZJRSITC-UHFFFAOYSA-M
sodium;(4-amino-1-hydroxy-1-phosphonobutyl)-hydroxyphosphinate;trihydrate
23681107
271.08
Crystalline Powder

Chemical Identifiers

121268-17-5
325.122
DCSBSVSZJRSITC-UHFFFAOYSA-M
23681107
sodium;(4-amino-1-hydroxy-1-phosphonobutyl)-hydroxyphosphinate;trihydrate
C4H18NNaO10P2
MFCD01748233
alendronate sodium hydrate, alendronate sodium trihydrate, alendronate sodium, alendronate monosodium trihydrate, alendros, adronat, fosamax, sodium alendronate hydrate, elandor, fosalan
CHEBI:2566
C(CC(O)(P(=O)(O)O)P(=O)(O)[O-])CN.O.O.O.[Na+]
Videos
Safety and Handling

Safety and Handling

RTECSNumber : SZ6523666

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions