Abbott™ ID NOW Influenza A and B 2 DFS Item
The fastest molecular flu test just got faster and even better. Now with early detection and positive results in as few as 5 minutes, allowing you to make effective clinical decisions sooner. Abbott ID NOW is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.
Supplier:  Abbott™ 427000
Catalog No. 23-046-634
Explore available promotions
Description
Description
 Transforming near-patient testing
  • Unique isothermal amplification technology giving you molecular results, faster than ever before
Improving patient outcomes
  • Giving you the confidence to make effective clinical decisions sooner
Optimizing operational efficiency
  • Enables prompt initiation of infection control measures
  • Connected results direct to your network
  • Facilitating effective patient management
Enabling effective Antimicrobial Stewardship
  • Molecular sensitivity in an actionable timeframe allows the prudent use of antibiotics and antivirals
A positive impact on your facility
  • Reliable near-patient testing reduces overall healthcare costs
Confidence at the point of care
  • Intuitive and reliable, allowing you to respond quickly and accurately in any setting
  • Simple and easy to use molecular diagnostic test
  • Sample type nasal swab
  • Requires only minimal training
  • Small footprint can be used in any healthcare setting
  • Streamlined workflow connectivity can be provided to your network
  • Demonstrates impressive sensitivity for the detection of both influenza A and B in clinical respiratory specimens
Specifications
Specifications
CLIA waived
Detects up to 42% more true positives than RADTs
Influenza testing
Room Temperature
Abbott ID NOW Influenza A and B Test Kit