missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Molecular Probes™ Alexa Fluor™ 350 Antibody Labeling Kit DFS Item

This kit is optimized for labeling 100 μg of antibody per reaction with blue fluorescent Alexa Fluor™ 350.

Supplier:  Molecular Probes™ A20180

Catalog No. A20180

Explore available promotions

Description

Description

Molecular Probes™ Alexa Fluor™ Antibody Labeling Kits provide a convenient means to label small amounts of antibodies with Alexa Fluor™ dyes (choice of 10 colors). This kit is optimized for labeling 100μg of antibody per reaction with blue fluorescent Alexa Fluor™ 350. Comparably small amounts of other proteins (>40 kDa) can also be labeled.

The kit contains everything you need to perform five separate labeling reactions as well as to purify the resulting conjugates. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry, primary detection, ELISAs, immunocytochemistry, indirect FISH, and more.

Important Features of Alexa Fluor™ 350 Antibody Labeling Kit:
• With an excitation and emission maximum of 346/442 nm, Alexa Fluor™ 350 can be efficiently excited using a UV laser line and detected under standard DAPI filters
• Labeled proteins typically ready to use typically in 90 min (∼15 min hands-on time)
• Useful for labeling 100μg of protein
• Optimized for small-scale labeling of any protein >40 kDa
• Purified using convenient spin filters
• Stabilizing proteins must be removed from the sample before labeling
• Includes detailed instructions for determining degree of labeling (DOL)

Better Results and Workflows with Primary labeled antibodies
A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Further, multiple primary antibodies of the same isotype or derived from the same species can easily be used in the same experiment if they are directly labeled with compatible fluorophores.

Superior Alexa Fluor™ Dyes
Compared to traditional dyes, Alexa Fluor™ dyes are brighter, more photostable, and more pH resistant between pH 4 and 10. And generally when using Alexa Fluor™ dyes, higher degrees of labeling can be achieved without intramolecular quenching. For details see Alexa Fluor™ Dyes Spanning the Visible and Infrared Spectrum—Section 1.3.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.

We'll Make a Custom Antibody Conjugate for You
If you can't find what you're looking for in our stocked list, we'll prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Specifications

Specifications

Labeling Kit
Alexa Fluor™ 350 Antibody Labeling Kit comes with 5 vials of reactive dye, sodium bicarbonate, purification resin, spin columns and collections tubes.
  • Store at 2°C to 8°C, protected from light. Component A may be stored from -20°C to +8°C.
  • 346/442 nm
    100 μg
    Alexa Fluor™
    Blue
    Alexa Fluor
    1 Kit
    Fluorescence
    Conjugation-based
    Room Temperature
    Alexa Fluor 350
    Antibodies, Proteins
    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions

    For Research Use Only. Not for use in diagnostic procedures.