missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlgiMatrix™ 6-Well, 3D Culture System, Flat-Bottom Microplate DFS Item

Color: Clear; Sterility: Sterile; Lid: With Lid

Supplier:  Gibco™ A1098201

 View more versions of this product

Catalog No. A1098201


Only null left
Explore available promotions

Description

Description

AlgiMatrix™ 3D Culture System 6 well plate is a user-friendly, animal –free bioscaffold available for the development of high-fidelity cell culture models that are more predictive of disease states and drug responses. AlgiMatrix™ sponges are formulated using pharmaceutical-grade raw material from brown seaweed. Its pure, chemically defined, and nontoxic macro porous structure improved lot-to-lot consistency, superior cell loading and excellent nutrient delivery, without damage to cells. The AlgiMatrix™ sponge is biodegradable and is stable at room temperature.
 
The AlgiMatrix™ 3D Culture System is suitable for a broad range of procedures; including Multicellular tumor spheroid assays (MCTS), hepatocyte and cardiomyocyte organogenesis studies, co-culture studies, and high-throughput drug screening assays.
Specifications

Specifications

With Lid
Algimatrix
Microplate
Sterile
Flat Bottom (F-well)
Cell Culture Grade
Room Temperature
Contains 1 x 6 well AlgiMatrix™ plate and 50 ml firming buffer. Store at room temperature, 15-30° C
Clear
6
AlgiMatrix 3D Culture System
1 x 6 well AlgiMatrix Plate and 50 mL Firming Buffer
1 x 6 well plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.