missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alginic acid sodium salt, low viscosity, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2526630

 View more versions of this product

Catalog No. AAB2526630


Explore available promotions

Description

Description

A gelling polysaccharide extracted from giant brown seaweed, it is useful in viscosity studiesAlginic acid sodium salt is widely utilized in the food industry as flavorless gum which enhances viscosity and acts as an emulsifier. It finds application as a thickener for reactive dyestuffs which is used in textile screen printing and carpet jet-printing. It is used as indigestion tablets and for the preparation of dental impressions. As a polymer, it combines with chitosan to synthesize a biodegradable porous scaffold which is useful for tissue engineering applications. It acts as a food additive and is used in production of gel-like foods such as baking. It is also used for encapsulating agents of microparticles of beta-galactosidae and in the preparation of alginate hydrogels with tunable mechanical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Alginic acid sodium salt
9005-38-3
MFCD00081310
Algin; Polymannuronic acid sodium salt
250 g
low viscosity
(C6H7O7)A(C6H7O7)BNa
14,241
Soluble in water. Insoluble in alcohol,chloroform and ether.
Hygroscopic

Chemical Identifiers

9005-38-3
(C6H7O7)A(C6H7O7)BNa
MFCD00081310
Algin; Polymannuronic acid sodium salt
Videos
Safety and Handling

Safety and Handling

H500

RTECSNumber : AZ5820000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only