missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aliquat∣r 336 TG, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A17247AE

 View more versions of this product

Catalog No. AAA17247AE


Explore available promotions

Description

Description

Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.

Solubility
Miscible with benzene and chloroform.

Notes
Incompatible with strong oxidizing agents, acids and bases.
Specifications

Specifications

Aliquat 336
0.88
132°C (269°F)
1.4465
100 mL
Hygroscopic
Miscible with benzene and chloroform.
CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.[Cl-]
404.164
CHEBI:75286
63393-96-4
>240°C
C25H54ClN
MFCD00011862
UN2810
methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride
XKBGEWXEAPTVCK-UHFFFAOYSA-M
methyl(trioctyl)azanium;chloride
21218
404.17

Chemical Identifiers

63393-96-4
MFCD00011862
methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride
CHEBI:75286
C25H54ClN
XKBGEWXEAPTVCK-UHFFFAOYSA-M
21218
methyl(trioctyl)azanium;chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314-H335-H360FD-H373

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 264-120-7

RTECSNumber : BR8575000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only