missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alizarin Red S (Sodium Sulfonate), MP Biomedicals™
Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage
Supplier: MP Biomedicals Inc 0210037525
Specifications
Alizarin Red S | |
130-22-3 | |
MFCD00013049 | |
HFVAFDPGUJEFBQ-UHFFFAOYSA-M | |
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate | |
3955344 | |
342.3 | |
25 g |
Indicator: pH Range 3.7 - 5.2 | |
C14H7NaO7S | |
Sodium Alizarinsulfonate, Alizarin sodium sulfonate | |
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+] | |
342.253 | |
CHEBI:87358 | |
Orange | |
Powder |
Chemical Identifiers
130-22-3 | |
342.253 | |
HFVAFDPGUJEFBQ-UHFFFAOYSA-M | |
3955344 | |
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate |
C14H7NaO7S | |
MFCD00013049 | |
Sodium Alizarinsulfonate, Alizarin sodium sulfonate | |
CHEBI:87358 | |
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+] |
Safety and Handling
EINECSNumber : 204-981-8
Recommended Storage : Store at Room Temperature (15-30°C).