missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alkaline Batteries
For peak performance in high-tech electronic devices and high-use devices
$14.03 - $14.03
Specifications
Product Type | Alkaline Battery |
---|---|
For Use With (Application) | For peak performance in high-tech electronic devices and high-use devices |
For Use With (Equipment) | For MP3 players, disc players, cameras, handheld TVs, handheld games, stereos, remote controlled devices |
Catalog Number | Mfr. No. | Battery Type | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Battery Type | Quantity | Price | Quantity | |||||
S43921
|
Customer Channels Group (Ccg)
EPD8154 |
AA | 4/Pk. |
Pack of 4 Each for $14.03
Case of 48 Pack for $673.44
|
|
|||||
Description
Use alkaline batteries for high-use applications like:
- MP3 Players
- Disc Players
- Cameras
- Handheld TVs
- Handheld Games
- Stereos
- Remote Controlled Devices
Specifications
Alkaline Battery | |
For peak performance in high-tech electronic devices and high-use devices | |
For MP3 players, disc players, cameras, handheld TVs, handheld games, stereos, remote controlled devices |