missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Coast Alkaline Battery 9 V Industrial Performance

Designed to maximize the performance of all types of handheld/headlamp devices, COAST Industrial Performance (IP) 9V batteries deliver top-rated power7mdash; whether you’re on the worksite, at home, or on the go

$20.89 - $244.64

Specifications

Battery Type 9 V
Cell Type Button
Voltage 9 V
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
17-018-113
View Documents
Coast
30602
4/Pk.
Pack of 4 Each for $20.89
 
17-018-044
View Documents
Coast
30603
12/Pk.
Pack of 12 Each for $62.07
 
17-004-841
View Documents
Coast
30604
48/Pk.
Pack of 48 Each for $244.64
 
Description

Description

  • Remove the stress from critical situations and simplify routine, practical situations by choosing batteries that take performance to the next level
  • When you choose COAST Industrial Performance batteries, you get the quality guarantee of a company that’s been at it for 100 years—with products that make people’s work safer and lives easier
Specifications

Specifications

9 V
Button
9 V
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'