missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Metro™ All Stainless Steel Carts for Labs and Cleanrooms
All Stainless Steel Carts for Labs and Cleanrooms are designed to handle regular cleaning and sterilization without compromising its structural integrity.
Supplier: Metro™ CRLS223NFS
Description
- Type 304 stainless steel construction
- All stainless steel shelves and corners
- Stainless steel posts
- Metro 5MPGSA/5MPBGSA 5 in. polyurethane cart-washable stem casters (2 swivel/2 brake)
Specifications
Cleanroom Utility Cart | |
Silver | |
900 lb. | |
Stainless Steel |
408 kg | |
34.75 x 18 x 39 in. | |
408 kg | |
3 |